Learn More
CPC Scientific H-Ser-Phe-Phe-Leu-Arg-Asn-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific PARP003A
Encompass
PAR-1 agonist.ONE-LETTER SEQUENCE: SFFLRNMOLECULAR FORMULA: C37H54N10O9MOLECULAR WEIGHT:782.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [140436-67-5], RESEARCH AREA: HematologyREFERENCES: Z.Suo et al., J. Biol. Chem., 278, 37681 (2003); P.J.Yu et al., Oncogene, 27, 2594 (2008); F.Bock et al., Proc. Natl. Acad. Sci. USA, 110, 648 (2013)
Catalog No. 50-211-0098