Learn More
CPC Scientific H-Thr-Phe-Leu-Leu-Arg-Asn-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PARP024A
Encompass
This peptide is a PAR-1 selective peptide induces cyclooxygenase-2 (COX-2) protein and mRNA expression in human endothelial cells. This peptide increases lung edema during high-tidal-volume ventilationSEQUENCE: H-Thr-Phe-Leu-Leu-Arg-Asn-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: TFLLRNMOLECULAR FORMULA: C35H58N10O9MOLECULAR WEIGHT: 762.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: HematologyREFERENCES:Syeda,, F. et al. J. Biol. Chem. 281, 11792 (2006)Jenkins, R. et al. J. Clin. Invest., 116, 1606 (2006)SKU(s): PARP-024A
Catalog No. 50-211-0137