Learn More
Bachem PAR-2 (1-6) amide (human) Trifluoroacetate
SDP
Supplier:  Bachem 4029152.0025
Encompass_Preferred
PAR-2 (1-6) amide (human) Trifluoroacetate, H-Ser-Leu-Ile-Gly-Lys-Val-NH2, 190383-13-2, C28H54N8O7, Mr 614.79, 25mg. Thrombin Receptor-Like 1 (1-6) amide (human), Proteinase Activated Receptor 2 (1-6) amide (human), Coagulation Factor II Receptor-Like 1 (1-6) amide (human), SLIGKVamide. SLIGKV-amide, which corresponds to the tethered ligand sequence of human PAR-2 (protease activated receptor 2), can be used to investigate receptor functions.
Catalog No. 50-259-860