Learn More
CPC Scientific H-Ser-Leu-Ile-Gly-Arg-Leu-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific PARP006B
Encompass
The peptide is a protease-activated receptor-2 (PAR2) selective agonist.SEQUENCE: H-Ser-Leu-Ile-Gly-Arg-Leu-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: SLIGRL-NH2MOLECULAR FORMULA: C29H56N10O7MOLECULAR WEIGHT: 656.83STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [171436-38-7]SYNONYMS: Proteinase Activated Receptor 2 (1-6) amide (mouse, rat), SLIGRLamide, Thrombin Receptor-Like 1 (1-6) amide (mouse, rat), Coagulation Factor II Receptor-Like 1 (1-6) amide (mouse, rat), SLIGRL-NH2 Protease-Activated Receptor 2 (PAR2) AgonistRESEARCH AREA: HematologyREFERENCES:B.al-Ani et al., Can. J. Physiol. Pharmacol., 73, 1203 (1995)M.D.Hollenberg et al., Mol. Pharmacol., 49, 229 (1996)M.Saiffeddine et al., Br. J. Pharmacol., 118, 521 (1996)N.Vergnolle, J. Immunol., 163, 5064 (1999)B.Al-Ani et al., Br. J. Pharmacol., 128, 1105 (1999)E.R.Sharlow et al., J. Cell Sci., 113, 3093 (2000)SKU(s): PARP-006A, PARP-006B
Catalog No. 50-211-0105