Learn More
CPC Scientific H-Leu-Ser-Ile-Gly-Lys-Val-NH2 (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific PARP001C
Encompass
This peptide is a partially reversed human PAR-2 activating peptide serving as an inactive control for the PAR-2 agonist SLIGKV-amide.ONE-LETTER SEQUENCE: LSIGKV-NH2MOLECULAR FORMULA: C28H54N8O7MOLECULAR WEIGHT:614.79STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1348395-60-7], SYNONYMS: LSIGKVamideRESEARCH AREA: HematologyREFERENCES: H.Nishikawa et al., J. Pharmacol. Exp. Ther., 312, 324 (2005); I.A. Akers et al.,Am. J. Physiol. Lung Cell. Mol. Physiol., 278, L193 (2000); S. Miyata, et al., J. Biol. Chem., 275, 4592 (2000); S. Miike, A.S. McWilliam, and H. Kita, J. Immunol., 167, 6615 (2001)
Catalog No. 50-211-0094