Learn More
Medchemexpress LLC PAR-2-IN-1 | 1690176-75-0 | 98.9% | 267.71 | 50 MG
SDP
Catalog No. 5000348812
Encompass_Preferred
PAR-2-IN-1 is a protease-activated receptor-2 (PAR2) signaling pathway inhibitor. It demonstrates anti-inflammatory and anticancer effects.
  • Protease-activated receptor-2 (PAR2) signaling pathway inhibitor
  • Exhibits anti-inflammatory effects
  • Demonstrates anticancer effects
  • Related to cancer targeted therapy and cancer metabolism
  • Acts on GPCR/G protein and protease activated receptor (PAR) pathways
Catalog No. 50-003-48812 Supplier Medchemexpress LLC Supplier No. HY13855850MG
PAR-2-IN-1 is a protease-activated receptor-2 (PAR2) signaling pathway inhibitor. It demonstrates anti-inflammatory and anticancer effects.
  • Protease-activated receptor-2 (PAR2) signaling pathway inhibitor
  • Exhibits anti-inflammatory effects
  • Demonstrates anticancer effects
  • Related to cancer targeted therapy and cancer metabolism
  • Acts on GPCR/G protein and protease activated receptor (PAR) pathways