Learn More
CPC Scientific H-Thr-Phe-Arg-Gly-Ala-Pro-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific PARP007A
Encompass
This peptide mimicks the putative tethered ligand of human PAR-3 (protease activated receptor 3). PAR3 Tethered Ligand (Human) / Activates PAR1 and PAR2SEQUENCE: H-Thr-Phe-Arg-Gly-Ala-Pro-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: TFRGAPMOLECULAR FORMULA: C29H45N9O8MOLECULAR WEIGHT: 647.73STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [320347-28-2]SYNONYMS: PAR3 (1-6) (human), Proteinase Activated Receptor 3 (1-6) (human), Thrombin Receptor-Like 2 (1-6) (human), Coagulation Factor II Receptor-Like 2 (1-6) (human)RESEARCH AREA: HematologyREFERENCES:H.Ishihara et al., Nature, 386, 502 (1997)K.K. Hansen et al., Immunology, 112, 183 (2004)SKU(s): PARP-007A
Catalog No. 50-211-0106