Learn More
CPC Scientific H-Ala-Tyr-Pro-Gly-Lys-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PARP012A
Encompass
SEQUENCE: H-Ala-Tyr-Pro-Gly-Lys-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: AYPGKFMOLECULAR FORMULA: C34H47N7O8MOLECULAR WEIGHT: 681.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [380900-00-5]SYNONYMS: (Ala)-Proteinase Activated Receptor 4 (1-6) amide (mouse), AYPGKFamide, (Ala)-Thrombin Receptor-Like 3 (1-6) amide (mouse), (Ala)-Coagulation Factor II Receptor-Like 3 (1-6) amide (mouse), AYPGKF-NH2 Selective Protease-Activated Receptor 4 (PAR4) AgonistRESEARCH AREA: Hematology, CardiovascularREFERENCES:P.Andr et al., J. Clin. Invest., 112, 398 (2003)D.Woulfe et al., J. Clin. Invest., 113, 441 (2004)E.A. Lidington, et al., Am J Physiol Cell Physiol, 289, C1437 (2005)M.D. Hollenberg, et al., Pharmacol., 143, 443 (2004)SKU(s): PARP-012A, PARP-012B
Catalog No. 50-211-0114