Learn More
Genscript Corporation PAR-4 Agonist Peptide, amide
Supplier:  Genscript Corporation RP115295
Encompass
PAR-4 agonist peptide stimulates thromboxane production by human platelets with the maximal response to this agonist being approximately half of that observed after maximal thrombin stimulation.
Catalog No. 50-275-102