Learn More
CPC Scientific H-Gly-Tyr-Pro-Gly-Gln-Val-NH2
(trifluoroacetate salt) 5MG
SDP
Catalog No. 502110116
Encompass
Tethered ligand sequence of human PAR-4.SEQUENCE: H-Gly-Tyr-Pro-Gly-Gln-Val-NH2(trifluoroacetate salt)ONE-LETTER SEQUENCE: GYPGQV-NH2MOLECULAR FORMULA: C28H42N8O8MOLECULAR WEIGHT: 618.69STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [245443-51-0]SYNONYMS: Proteinase Activated Receptor 4 (1-6) amide (human), GYPGQVamide, Coagulation Factor II Receptor-Like 3 (1-6) amide (human), Thrombin Receptor-Like 3 (1-6) amide (human), GYPGQV-NH2 Selective Protease-Activated Receptor 4 (PAR4) AgonistRESEARCH AREA: HematologyREFERENCES:M.D.Hollenberg et al., Can. J. Physiol. Pharmacol., 77, 458 (1999)M.Tognetto et al., Br. J. Pharmacol., 139, 21 (2003)M.D.Hollenberg and S.Houle, Swiss Med. Wkly., 135, 425 (2005)SKU(s): PARP-013A, PARP-013B
Catalog No. 50-211-0116 Supplier CPC Scientific Supplier No. PARP013A
Tethered ligand sequence of human PAR-4.SEQUENCE: H-Gly-Tyr-Pro-Gly-Gln-Val-NH2(trifluoroacetate salt)ONE-LETTER SEQUENCE: GYPGQV-NH2MOLECULAR FORMULA: C28H42N8O8MOLECULAR WEIGHT: 618.69STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [245443-51-0]SYNONYMS: Proteinase Activated Receptor 4 (1-6) amide (human), GYPGQVamide, Coagulation Factor II Receptor-Like 3 (1-6) amide (human), Thrombin Receptor-Like 3 (1-6) amide (human), GYPGQV-NH2 Selective Protease-Activated Receptor 4 (PAR4) AgonistRESEARCH AREA: HematologyREFERENCES:M.D.Hollenberg et al., Can. J. Physiol. Pharmacol., 77, 458 (1999)M.Tognetto et al., Br. J. Pharmacol., 139, 21 (2003)M.D.Hollenberg and S.Houle, Swiss Med. Wkly., 135, 425 (2005)SKU(s): PARP-013A, PARP-013B