Learn More
Eurofins Discoverx ChemiScreen PAR2 Receptor Membrane Preparation, 1 vial. (HTS037M)
Catalog No. 502005719
Encompass
The ChemiScreen PAR2 Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of F2RL1 surface expression. ChemiScreen membrane preps can be used to screen for GPCR activity using GTPγS assays. Increased GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5719 Supplier Eurofins Discoverx Supplier No. HTS037M
The ChemiScreen PAR2 Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of F2RL1 surface expression. ChemiScreen membrane preps can be used to screen for GPCR activity using GTPγS assays. Increased GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.