Learn More
Cayman Chemical PAR2 (1-6) amide (human) (trifluoroacetate salt), Purity: ≥95%, CAS: 2379569-17-0, 50 mg
SDP
Catalog No. 502910687
Encompass_Preferred
A peptide agonist of PAR2; binds to NCTC 2544 cells expressing human PAR2 (Ki = 9.64 µM in a radioligand binding assay) and induces calcium mobilization in the same cells (EC50 = 0.075 µM); reduces colony formation of A549 cells at 100 µM; induces superoxide production and degranulation in isolated human eosinophils at 500 µM; induces tear secretion in rats at 5 µmol/kg when used in combination with amastatin
Catalog No. 50-291-0687 Supplier Cayman Chemical Supplier No. 3520450 MG
A peptide agonist of PAR2; binds to NCTC 2544 cells expressing human PAR2 (Ki = 9.64 µM in a radioligand binding assay) and induces calcium mobilization in the same cells (EC50 = 0.075 µM); reduces colony formation of A549 cells at 100 µM; induces superoxide production and degranulation in isolated human eosinophils at 500 µM; induces tear secretion in rats at 5 µmol/kg when used in combination with amastatin