Learn More
Cayman Chemical PAR41-6humn 10mg
SDP
Catalog No. 502937239
Encompass_Preferred
A peptide agonist of PAR4; activates PAR4 and the cleavage site mutant PAR4R47A at 500 µM; induces platelet aggregation of isolated washed human platelets at 1 mM but does not affect clotting time ex vivo
Catalog No. 50-293-7239 Supplier Cayman Chemical Supplier No. 2712610 MG
A peptide agonist of PAR4; activates PAR4 and the cleavage site mutant PAR4R47A at 500 µM; induces platelet aggregation of isolated washed human platelets at 1 mM but does not affect clotting time ex vivo