Learn More
Cayman Chemical PAR4 (1-6) (mouse) (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502938828
Encompass_Preferred
A peptide PAR4 agonist (EC50 = 30 μM in Xenopus oocytes expressing human PAR4 receptors); corresponds to residues 1-6 of the amino terminal tethered ligand sequence of mouse PAR4 and residues 60-65 of the full-length sequence; induces calcium release in wild-type and PAR3-/- mouse platelets and in human platelets; triggers [3H]inositol phosphate release from KOLF cells expressing human PAR4
Catalog No. 50-293-8828 Supplier Cayman Chemical Supplier No. 2713110 MG
A peptide PAR4 agonist (EC50 = 30 μM in Xenopus oocytes expressing human PAR4 receptors); corresponds to residues 1-6 of the amino terminal tethered ligand sequence of mouse PAR4 and residues 60-65 of the full-length sequence; induces calcium release in wild-type and PAR3-/- mouse platelets and in human platelets; triggers [3H]inositol phosphate release from KOLF cells expressing human PAR4