Learn More
IMEB, Inc IMEB Para-Ban Paraffin Cleaner/Repellent, 4oz, 6/CS
SDP
Catalog No. 501689477
Encompass
The world’s finest paraffin repellent, as the name implies PARA-BAN will help keep your microtome free of paraffin. Just spray it on any surface where paraffin normally sticks. Good for table tops and most microtomes. PARA-BAN is safe on any metal or plastic surface. Used by histologists everywhere. Cleans and repels in one simple spray. Apply with paper towel or gauze pad to desired surface.
Catalog No. 50-168-9477 Supplier IMEB, Inc Supplier No. PB604
The world’s finest paraffin repellent, as the name implies PARA-BAN will help keep your microtome free of paraffin. Just spray it on any surface where paraffin normally sticks. Good for table tops and most microtomes. PARA-BAN is safe on any metal or plastic surface. Used by histologists everywhere. Cleans and repels in one simple spray. Apply with paper towel or gauze pad to desired surface.