Learn More
Cayman Chemical para-bromo 4-ANPP, Purity: ≥98%, CAS: 2752105-11-4, 1 mg
SDP
Catalog No. 502937339
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids; a precursor in the synthesis of para-bromofentanyl; intended for research and forensic applications
Catalog No. 50-293-7339 Supplier Cayman Chemical Supplier No. 282161 MG
An analytical reference standard that is structurally similar to known opioids; a precursor in the synthesis of para-bromofentanyl; intended for research and forensic applications