Learn More
Cayman Chemical para-bromo Methylaminorex, Purity: ≥98% (mixture of diastereomers), 5 mg
SDP
Catalog No. 502946582
Encompass_Preferred
An analytical reference standard that is structurally similar to known stimulants; intended for research and forensic applications
Catalog No. 50-294-6582 Supplier Cayman Chemical Supplier No. 328355 MG
An analytical reference standard that is structurally similar to known stimulants; intended for research and forensic applications