Learn More
Cayman Chemical para-fluoro Phenethyl 4-ANPP (hydrochloride), Purity: ≥98%, CAS: 2840954-90-5, 5 mg
SDP
Catalog No. 502948628
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications
Catalog No. 50-294-8628 Supplier Cayman Chemical Supplier No. 335655 MG
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications