Description
Achieve faster detection of M. avium ssp. paratuberculosis with the Thermo Scientific™ VersaTREK™ System and Thermo Scientific™ para-JEM™ reagents, including para-JEM Broth.
The para-JEM System includes a number of unique features:
- Detects faster—able to recover MAP in as little as 2 to 3 weeks, much earlier than conventional methods*
- Provides increased sensitivity over conventional methods*
- Provides semi-quantitation for low, medium and high shedders based on time-to-detection*
- Reduces labor with fewer false positives, false negatives and contaminants*
- Improves workflow with most tests concluded at six weeks for high, medium and many low shedders*
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | para-JEM Broth |
| Product Type | Microbiology Media |
| Test Type | Microbial Detection |
| No. per Pack | 50 |
| Packaging Type | 10.5 mL Each |
| Quantity | 10.5 mL |
| Specimen Type | Various |
| Species | M. avium ssp. paratuberculosis |