Learn More
Thermo Scientific™ para-JEM™ EYS
GSA_VA
Catalog No. Y713442 Shop All Thermo Scientific Products
Change view
Click to view available options
Packaging Type:
50 mL Each
Quantity:
50 mL
Catalog No. Packaging Type Quantity
Y713442 50 mL Each 50 mL
1 options
This item is not returnable. View return policy
Catalog No. Y713442 Supplier Thermo Scientific™ Supplier No. Y713442
Add to Cart
Add to Cart
This item is not returnable. View return policy
Facilitate faster detection of M. aviumssp. paratuberculosis with Thermo Scientific™ para-JEM™ EYS egg yolk supplement.
Protect slow growing mycobacteria and facilitate faster detection of M. avium ssp. paratuberculosis with Thermo Scientific™ para-JEM™ EYS egg yolk supplement.
Solid medium culture used to be the standard procedure for detecting M. avium ssp. paratuberculosis (MAP), the causative agent of Johne's Disease, in ruminant fecal samples before para-JEM reagents. Due to the bacterium's long generation time, it can take up to 12 weeks of incubation on solid medium before the bacteria are detected. The para-JEM System is a unique, liquid culture method that combines a full line of reagents with the powerful Thermo Scientific™ VersaTREK™ Automated Microbial Detection System for faster, automated detection of MAP.
The para-JEM System includes a number of unique features:
  • Detects faster—able to recover MAP in as little as 2 to 3 weeks, much earlier than conventional methods*
  • Provides increased sensitivity over conventional methods*
  • Provides semi-quantitation for low, medium and high shedders based on time-to-detection*
  • Reduces labor with fewer false positives, false negatives and contaminants*
  • Improves workflow with most tests concluded at six weeks for high, medium and many low shedders* 
Utilize para-JEM reagents with the fully automated VersaTREK Microbial Detection System, which automatically incubates and continuously monitors culture bottles for accurate, reliable organism detection as well as enhanced ergonomics and workflow versatility.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description para-JEM EYS
Product Type para-JEM Growth Supplement
Test Type Microbial Detection
No. per Pack 5
Packaging Type 50 mL Each
Quantity 50 mL
Specimen Type Various
Species M. avium ssp. paratuberculosis