Learn More
Electron Microscopy Sciences Para-Marker
SDP
Catalog No. 50930982
Encompass_Preferred
A wide tipped pen, designed for use with paraffin sections. The pen provides a sticky membrane to the glass slide which not only prevents paraffin sections from falling off but also keeps the sections flat on the slide. A wide tipped pen designed for use with paraffin sections. The pen provides a sticky membrane to the glass slide which not only prevents paraffin sections from falling off but also keeps the sections flat on the slide.
Catalog No. 50-930-982 Supplier Electron Microscopy Sciences Supplier No. 7131320
A wide tipped pen, designed for use with paraffin sections. The pen provides a sticky membrane to the glass slide which not only prevents paraffin sections from falling off but also keeps the sections flat on the slide. A wide tipped pen designed for use with paraffin sections. The pen provides a sticky membrane to the glass slide which not only prevents paraffin sections from falling off but also keeps the sections flat on the slide.