Learn More
Cayman Chemical para-methoxy 4-ANPP, Purity: ≥98%, CAS: 28456-24-8, 5 mg
SDP
Catalog No. 502940822
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids; a precursor in the synthesis of para-methoxyfentanyl; a potential impurity in para-methoxyfentanyl preparations; intended for research and forensic applications
Catalog No. 50-294-0822 Supplier Cayman Chemical Supplier No. 279465 MG
An analytical reference standard that is structurally similar to known opioids; a precursor in the synthesis of para-methoxyfentanyl; a potential impurity in para-methoxyfentanyl preparations; intended for research and forensic applications