Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Para-nitrophenyl oleate | 17363-90-5 | 99.8% | 403.55 | 5 MG
SDP

Catalog No. 5000394488
Encompass_Preferred

Para-nitrophenyl oleate is an ester product. It is for research use only and not sold to patients.

  • Purity: 99.81%
  • Molecular weight: 403.55
  • Formula: C24H37NO4
  • CAS number: 17363-90-5
  • Appearance: Liquid (Density: 1.017±0.06 g/cm3)
  • Color: Colorless to light yellow
  • Storage: -20°C for 3 years (pure form); -80°C for 6 months, -20°C for 1 month (in solvent)
  • Solubility in DMSO: 200 mg/mL (495.60 mM)

Catalog No. 50-003-94488 Supplier Medchemexpress LLC Supplier No. HY1658365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Para-nitrophenyl oleate is an ester product. It is for research use only and not sold to patients.

  • Purity: 99.81%
  • Molecular weight: 403.55
  • Formula: C24H37NO4
  • CAS number: 17363-90-5
  • Appearance: Liquid (Density: 1.017±0.06 g/cm3)
  • Color: Colorless to light yellow
  • Storage: -20°C for 3 years (pure form); -80°C for 6 months, -20°C for 1 month (in solvent)
  • Solubility in DMSO: 200 mg/mL (495.60 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.