Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Para-nitrophenyl oleate | 17363-90-5 | 99.8% | 403.55 | 50 MG
SDP

Catalog No. 5000394487
Encompass_Preferred

Para-nitrophenyl oleate is an ester product intended for research use only. This product features:

  • High purity of 99.81%
  • Molecular weight of 403.55
  • Chemical formula C24H37NO4
  • CAS Number 17363-90-5
  • Colorless to light yellow liquid appearance

Catalog No. 50-003-94487 Supplier Medchemexpress LLC Supplier No. HY16583650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Para-nitrophenyl oleate is an ester product intended for research use only. This product features:

  • High purity of 99.81%
  • Molecular weight of 403.55
  • Chemical formula C24H37NO4
  • CAS Number 17363-90-5
  • Colorless to light yellow liquid appearance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.