Learn More
Medchemexpress LLC Para-nitrophenyl oleate | 17363-90-5 | 99.8% | 403.55 g/mol | C24H37NO4 | 10 MG
SDP
Catalog No. 5000289271
Encompass_Preferred
Para-nitrophenyl oleate is the p-nitrophenyl ester of oleic acid used as a chromogenic substrate in biochemical assays; hydrolysis releases p-nitrophenol that can be measured spectrophotometrically.
  • High purity (99.8%), appropriate for analytical and biochemical use.
  • Chromogenic substrate that produces a colorimetric readout on enzymatic hydrolysis.
  • Available in small research pack sizes suitable for assay development and screening.
  • Stable in pure form when stored at -20°C; reduced stability in solution.
  • Certificate of analysis and safety data sheet available for quality and handling information.
Catalog No. 50-002-89271 Supplier Medchemexpress LLC Supplier No. HY16583610MG
Para-nitrophenyl oleate is the p-nitrophenyl ester of oleic acid used as a chromogenic substrate in biochemical assays; hydrolysis releases p-nitrophenol that can be measured spectrophotometrically.
  • High purity (99.8%), appropriate for analytical and biochemical use.
  • Chromogenic substrate that produces a colorimetric readout on enzymatic hydrolysis.
  • Available in small research pack sizes suitable for assay development and screening.
  • Stable in pure form when stored at -20°C; reduced stability in solution.
  • Certificate of analysis and safety data sheet available for quality and handling information.