Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Para-Trim Plastic Paraffin Scraper
SDP

Catalog No. 22000074
Change view
Click to view available options
For Use With (Equipment):
Microtome
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-000-074 Microtome
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-074 Supplier StatLab Supplier No. PTSLMP
Add to Cart
Add to Cart

Helps remove dried paraffin and soft wax deposits from smooth surfaces

  • Removes dried paraffin
  • Removes soft wax deposits
  • Pink

Specifications

Product Type Paraffin Scraper
For Use With (Equipment) Microtome
Material Plastic
Disposable Reusable
Length (English) 0.122 in.
Length (Metric) 3.1 mm
Color Pink
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.