Learn More
ATCC Parabacteroides; distasonis
Supplier:  ATCC 8503
Encompass
Parabacteroides distasonis is a whole-genome sequenced bacterial type strain that produces superoxide dismutase and catalase. This culture can be used as a quality control strain for bioMerieux Vitek and IDS products and Remel RapID™ Systems.
Catalog No. 50-238-5045
 Restricted Item