Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paracetamol -D-glucuronide, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013281 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3281 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3281 Supplier MilliporeSigma Supelco Supplier No. 430735MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 16110-10-4
Physical Form Neat
Linear Formula C14H17 NO8
MDL Number MFCD03094680
Synonym p-Acetamidophenyl beta-D-glucuronide; Acetaminophen glucuronide
Type Analytical Standard
Quantity 5 mg
Formula Weight 327.29
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.