Learn More
Apacor ParaClick™ Pinworm Paddle Box (30/Pk.)
Catalog No. 22010107
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
22-010-107 Non-sterile
1 options
This item is not returnable. View return policy
Catalog No. 22-010-107 Supplier Apacor Supplier No. 250030
Add to Cart
Add to Cart
This item is not returnable. View return policy
ParaClick™ Pinworm Paddle Box for the detection of Enterobius Vermicularis Ova.
Intended use:
The ParaClick™ pinworm paddle is a sampling and microscopy device for the detection of enterobiasis ova to aid in the diagnosis of enterobiasis infection.
Features:
  • Suitable for qualitative verification
  • Economical and easy to use
  • Ready to use
  • Optimum sample clarity
  • Rapid 2 step process
  • Room temperature storage
  • 3 year shelf life
  • CE Marked
  • User friendly sample collection
Specifications
Product Type Pinworm Paddle
Format Paddle
Sterility Non-sterile