Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apacor ParaClick™ Patient Care (3 x 10/Pk.)

Catalog No. 22332105
Change view
Click to view available options
Quantity:
30/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-332-105 30/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-332-105 Supplier Apacor Supplier No. 250000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ParaClick™ Patient Care for the detection of Enterobius Vermicularis Ova.

Intended use:

The ParaClick™ pinworm paddle is a sampling and microscopy device for the detection of enterobiasis ova to aid in the diagnosis of enterobiasis infection.

Features:

  • Suitable for qualitative verification
  • Economical and easy to use
  • Ready to use
  • Optimum sample clarity
  • Rapid 2 step process
  • Room temperature storage
  • 3 year shelf life
  • CE Marked
  • User friendly sample collection

Specifications

Product Type Collection and Transport System
Format Swab
Sterility Non-sterile
Color Clear
For Use With (Application) Detection of Enterobius Vermicularis
Quantity 30/Pk.
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.