Learn More
Strem, An Ascensus Company CAS 23927-40-4. 1g. racemic-412-Dibromo[2.2]paracyclophane min. 95%. MFCD00184346
SDP
Catalog No. 502110689
Encompass
CAS 23927-40-4. 1g. racemic-412-Dibromo[2.2]paracyclophane min. 95%. MFCD00184346. Molecular Weight 366.09. Molecular Formula C16H14Br2. Color/form white to off-white pwdr. Strem 06-0460. http//www.strem.com/catalog/v/06-0460/
Catalog No. 50-211-0689 Supplier Strem, An Ascensus Company Supplier No. 0604601G
CAS 23927-40-4. 1g. racemic-412-Dibromo[2.2]paracyclophane min. 95%. MFCD00184346. Molecular Weight 366.09. Molecular Formula C16H14Br2. Color/form white to off-white pwdr. Strem 06-0460. http//www.strem.com/catalog/v/06-0460/