Learn More
Strem, An Ascensus Company CAS 612492-27-0. 500mg. racemic-412-Dihydroxy[2.2]paracyclophane min. 97%. MFCD08459337
SDP
Catalog No. 502110703
Encompass
CAS 612492-27-0. 500mg. racemic-412-Dihydroxy[2.2]paracyclophane min. 97%. MFCD08459337. Molecular Weight 240.30. Molecular Formula C16H16O2. Color/form yellow to brown pwdr. Strem 08-0700. http//www.strem.com/catalog/v/08-0700/
Catalog No. 50-211-0703 Supplier Strem, An Ascensus Company Supplier No. 080700500MG
CAS 612492-27-0. 500mg. racemic-412-Dihydroxy[2.2]paracyclophane min. 97%. MFCD08459337. Molecular Weight 240.30. Molecular Formula C16H16O2. Color/form yellow to brown pwdr. Strem 08-0700. http//www.strem.com/catalog/v/08-0700/