Learn More
IMEB, Inc Tek-Select Paraffin Block Metal Storage Cabinet 14 Drawers, Stackable
SDP
Catalog No. NC2598178
Encompass_Preferred
Tek-Select Paraffin Block Metal Storage Cabinet 14 Drawers, Stackable. Tek-Select economical light weight metal microscope slide and paraffin block storage cabinets allow for a secure and safe way of storing microscope slides and paraffin blocks conveniently together or stacked separately. Each metal file system is interlocking and can be stacked 12-15″ high. Cassette Storage Cabinet Stackable/Lockable for safety. Each unit contains 14 drawers.
Catalog No. NC2598178 Supplier IMEB, Inc Supplier No. SC801
Tek-Select Paraffin Block Metal Storage Cabinet 14 Drawers, Stackable. Tek-Select economical light weight metal microscope slide and paraffin block storage cabinets allow for a secure and safe way of storing microscope slides and paraffin blocks conveniently together or stacked separately. Each metal file system is interlocking and can be stacked 12-15″ high. Cassette Storage Cabinet Stackable/Lockable for safety. Each unit contains 14 drawers.