Learn More
Phoenix Metal Products Inc Tissue Block Storage Cabinet
SDP
Catalog No. NC1908196
Encompass
The TB-400 is for the storage of tissue blocks, also known as paraffin blocks, or can store tissue cartridges or embedding rings. This cabinet has been designed to be an economical yet durable method for storing large quantities of blocks. Priced just slightly higher per block than corrugated storage boxes, the TB-400 is a superior value as it will last a lifetime with its steel construction. Say goodbye to soggy, sagging and stale corrugated tissue block boxes, and say hello to the TB-400 tissue block storage cabinet.
Catalog No. NC1908196 Supplier Phoenix Metal Products Inc Supplier No. TB400
The TB-400 is for the storage of tissue blocks, also known as paraffin blocks, or can store tissue cartridges or embedding rings. This cabinet has been designed to be an economical yet durable method for storing large quantities of blocks. Priced just slightly higher per block than corrugated storage boxes, the TB-400 is a superior value as it will last a lifetime with its steel construction. Say goodbye to soggy, sagging and stale corrugated tissue block boxes, and say hello to the TB-400 tissue block storage cabinet.