Learn More
Med Vet International Paraffin Dispenser
Catalog No. 503043884
Encompass_Preferred
Large capacity (2.3 gallon) tabletop style dispenser. Industrial metal outer shell with durable stainless steel, easy to clean interior. Built in temperature controller and heated faucet to keep paraffin flowing. New lid with hole for thermometer! Temperature maximum: 72 degrees (tolerance ±2 degrees) Shipping weight: 46 lbs.
Catalog No. 50-304-3884 Supplier Med Vet International Supplier No. XH4002
Large capacity (2.3 gallon) tabletop style dispenser. Industrial metal outer shell with durable stainless steel, easy to clean interior. Built in temperature controller and heated faucet to keep paraffin flowing. New lid with hole for thermometer! Temperature maximum: 72 degrees (tolerance ±2 degrees) Shipping weight: 46 lbs.