Learn More
Leica Biosystems Paraffin Drain Hose Assembly for Leica Paraffin-Processing Equipment
Catalog No. NC1610314
Encompass
Replacement paraffin drain hose assembly designed for Leica Biosystems paraffin-processing instruments. Routes and disposes of molten paraffin and paraffin-contaminated fluids to support instrument cleanliness and reliable operation. Intended as a direct spare for routine maintenance and repairs; verify model compatibility before installation.
  • Provides a direct replacement for damaged or worn drain hoses.
  • Enables controlled routing and disposal of molten paraffin.
  • Supports routine maintenance and reduces instrument downtime.
  • Simple to install when used with compatible equipment.
  • Dimensions and material are not specified on the manufacturer product page.
Catalog No. NC1610314 Supplier Leica Biosystems Supplier No. 14047634721
Replacement paraffin drain hose assembly designed for Leica Biosystems paraffin-processing instruments. Routes and disposes of molten paraffin and paraffin-contaminated fluids to support instrument cleanliness and reliable operation. Intended as a direct spare for routine maintenance and repairs; verify model compatibility before installation.
  • Provides a direct replacement for damaged or worn drain hoses.
  • Enables controlled routing and disposal of molten paraffin.
  • Supports routine maintenance and reduces instrument downtime.
  • Simple to install when used with compatible equipment.
  • Dimensions and material are not specified on the manufacturer product page.