Learn More
Nacalai USA Inc Paraffin Liquid 500 mL, colorless, viscous, SP grade for Cell Culture, CAS RN 8012-95-1, boiling point 300 ℃ or more, flash point 230 ℃
Catalog No. NC1506764
Encompass
NACALAI USA INC Paraffin Liquid is a specially prepared reagent designed for critical applications in cell culture. This colorless, viscous liquid is supplied in a 500 mL volume and is recognized for its high purity and suitability for scientific research. This paraffin liquid is specifically graded for cell culture applications, ensuring a high level of quality suitable for sensitive biological systems.
  • The liquid is colorless and viscous, providing a stable and inert environment.
  • It has a high boiling point, contributing to its stability under various laboratory conditions.
  • The flash point of the product is 230 °C (o.c.), ensuring safe handling.
  • It is soluble in diethyl ether and chloroform.
  • Density at 20 °C is between 0.86 and 0.89 g/mL.
Catalog No. NC1506764 Supplier Nacalai USA Inc Supplier No. 2613785
NACALAI USA INC Paraffin Liquid is a specially prepared reagent designed for critical applications in cell culture. This colorless, viscous liquid is supplied in a 500 mL volume and is recognized for its high purity and suitability for scientific research. This paraffin liquid is specifically graded for cell culture applications, ensuring a high level of quality suitable for sensitive biological systems.
  • The liquid is colorless and viscous, providing a stable and inert environment.
  • It has a high boiling point, contributing to its stability under various laboratory conditions.
  • The flash point of the product is 230 °C (o.c.), ensuring safe handling.
  • It is soluble in diethyl ether and chloroform.
  • Density at 20 °C is between 0.86 and 0.89 g/mL.