Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Paraffin Pastilles

Catalog No. M1071642504 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 x 2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1071642504 4 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1071642504 Supplier MilliporeSigma™ Supplier No. 1071642504
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as histological embedding media

  • Used in clinical diagnostics
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 15° to 25°C
Packaging Type Plastic Bag
Quantity 4 x 2.5 kg
For Use With (Application) Histology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.