Learn More
Electron Microscopy Sciences EMS Replacement Paraffin Tapper, 22 x 22 mm, 1.5 mm thick
SDP
Catalog No. 501931614
Encompass_Preferred
Paraffin tappers hold the heat so the tissue stays flat in the mold and is embedded in the same plane for easier cutting. The paraffin tappers are helpful with multiple piece skins, large pieces of uterus, and even those multi-piece biopsies. They reduce re-cuts and re-embeds, as the tissue is perfectly embedded in the embedding mold the first time.
Size: 22 x 22 mm
Thickness: 1.5 mm
Catalog No. 50-193-1614 Supplier Electron Microscopy Sciences Supplier No. 6284204
Paraffin tappers hold the heat so the tissue stays flat in the mold and is embedded in the same plane for easier cutting. The paraffin tappers are helpful with multiple piece skins, large pieces of uterus, and even those multi-piece biopsies. They reduce re-cuts and re-embeds, as the tissue is perfectly embedded in the embedding mold the first time.
Size: 22 x 22 mm
Thickness: 1.5 mm