Learn More
Biochain Institute Inc FDA Standard Tissue Array, 1; slides/PK
SDP
Catalog No. 50181846
Encompass_Preferred
BioChain provides FDA Standard Frozen Tissue Array (TMA) which is designed in conformance with FDA guideline and meeting the requirements for IHC (Immunohistochemistry) and IVD (In vitro Diagnostic Devices) certification. This tissue array has two slides which contain 30 different human normal tissue types and 3 donors per tissue type. It also contains the 41 most popular human tumor tissue samples.
Catalog No. 50-181-846 Supplier Biochain Institute Inc Supplier No. T82347011
BioChain provides FDA Standard Frozen Tissue Array (TMA) which is designed in conformance with FDA guideline and meeting the requirements for IHC (Immunohistochemistry) and IVD (In vitro Diagnostic Devices) certification. This tissue array has two slides which contain 30 different human normal tissue types and 3 donors per tissue type. It also contains the 41 most popular human tumor tissue samples.