Learn More
Biochain Institute Inc Breast Tumor Tissue Array - duplicated 70 cases covering all the common types of breast cancer and 5 cases of normal and, 5 slides/PK
SDP
Catalog No. 50181858
Encompass_Preferred
BioChain's tissue arrays or tissue microarrays (TMAs) are a useful tool in biomarker research and drug target validation. It consists of paraffin blocks that are assembled in array fashion to allow multiplex histological analysis. BioChain's TMAs provide a valuable tool for analysis of protein, RNA, or DNA molecules in oncology, both the malignant and metastatic markers. TMAs can also been constructed with tissues from other diseases and species.
Catalog No. 50-181-858 Supplier Biochain Institute Inc Supplier No. Z7020004
BioChain's tissue arrays or tissue microarrays (TMAs) are a useful tool in biomarker research and drug target validation. It consists of paraffin blocks that are assembled in array fashion to allow multiplex histological analysis. BioChain's TMAs provide a valuable tool for analysis of protein, RNA, or DNA molecules in oncology, both the malignant and metastatic markers. TMAs can also been constructed with tissues from other diseases and species.