Learn More
Biochain Institute Inc Paraffin Tissue Section - Human Adult Normal: Brain: Cerebellar Peduncles, 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T2234038
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. BioChain's formalin fixed paraffin embedded (FFPE) tissue section line encompasses a large selection of tissue sections which are suitable for immunohistochemistry and in situ hybridization assays. The tissue sections are approximately 5 um thick. For those customers who want to perform FFPE DNA and RNA isolation, BioChain can provide tissue section slices in tubes instead of mounting them on glass slides.
Catalog No. 50-182-000