Learn More
Biochain Institute Inc Paraffin Tissue Section - Depression: Brain: Parietal Lobe, 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T2236066DEP
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. BioChain's formalin fixed paraffin embedded (FFPE) tissue section line encompasses a large selection of tissue sections which are suitable for immunohistochemistry and in situ hybridization assays. The tissue sections are approximately 5 um thick. Each slide contains tissue from single human diseased sample. Documentation of tissues' clinical histories may be available upon request. For those customers who want to perform FFPE DNA and RNA isolation, BioChain can provide tissue section slices in tubes instead of mounting them on glass slides.
Catalog No. 50-182-197