Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Human Tumor Same Tumor Type, Different Donors (5 tissues), 5 slides/PK
SDP
Catalog No. 50182538
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures.The tissues are immediately put in liquid nitrogen after being excised, and then identified by a board-licensed pathologist. Patient demographic and disease related history data are available. he tissue sections are approximately 5-10 um thick.
Catalog No. 50-182-538 Supplier Biochain Institute Inc Supplier No. T8235086
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures.The tissues are immediately put in liquid nitrogen after being excised, and then identified by a board-licensed pathologist. Patient demographic and disease related history data are available. he tissue sections are approximately 5-10 um thick.