Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Monkey (Cynomolgus) Normal Tissue, Multi-tissue I, 8 different tissues, 5 slides/PK
SDP
Catalog No. 50182549
Encompass_Preferred
BioChain's monkey normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different Cynomolgus monkey tissues. Tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-182-549 Supplier Biochain Institute Inc Supplier No. T8534448CY
BioChain's monkey normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different Cynomolgus monkey tissues. Tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.