Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Mouse Whole Brain Segmentation Panel, 5 slides/PK
SDP
Catalog No. 50182559
Encompass_Preferred
Whole brain segmentation panels are designed for study whole brain of small animals. The animals' whole brain was procured intact and was fixed in formaling, then it was sliced into 6 equal coronal regions. Each region is 2 mm thick. The 6 regions were embedded in the same paraffin block. The paraffin section panel cut from this block can enable scientists or research investigators do biochemical analysis, such as the determination of neurotransmitter and metabolite concentrations for reproducible brain regions (i.e. discrete terminal fields or nuclei).
Catalog No. 50-182-559 Supplier Biochain Institute Inc Supplier No. T8334035
Whole brain segmentation panels are designed for study whole brain of small animals. The animals' whole brain was procured intact and was fixed in formaling, then it was sliced into 6 equal coronal regions. Each region is 2 mm thick. The 6 regions were embedded in the same paraffin block. The paraffin section panel cut from this block can enable scientists or research investigators do biochemical analysis, such as the determination of neurotransmitter and metabolite concentrations for reproducible brain regions (i.e. discrete terminal fields or nuclei).