Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Human Adult Normal Tissue, Brain IV, 7 different tissues, 5 slides/PK
SDP
Catalog No. 50182511
Encompass_Preferred
BioChain's human adult normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different human normal tissues. Tissues mounted on positively charged glass slides are organized either by organs (major organ multiple tissue panels), or by integrated major systems (organ systems multiple tissue panels). Documentation of tissues' clinical histories is available upon request. Identical format total RNA Northern Blots are available, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-182-511 Supplier Biochain Institute Inc Supplier No. T8234564
Add to Cart
Add to Cart
BioChain's human adult normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different human normal tissues. Tissues mounted on positively charged glass slides are organized either by organs (major organ multiple tissue panels), or by integrated major systems (organ systems multiple tissue panels). Documentation of tissues' clinical histories is available upon request. Identical format total RNA Northern Blots are available, and cDNA from those tissues may also be available for PCR detection.