Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Monkey (Rhesus) Normal Tissue, Multi-tissue III, 5 slides/PK
SDP
Catalog No. 50182551
Encompass_Preferred
BioChain's monkey normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different Rhesus monkey tissues. Tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-182-551 Supplier Biochain Institute Inc Supplier No. T8534423
BioChain's monkey normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different Rhesus monkey tissues. Tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.