Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Human Disease Tissue, Alzheimer's Disease, Multi-tissue IV, 7 different tissues, 5 slides/PK
SDP
Catalog No. 50182522
Encompass_Preferred
BioChain's FFPE Alzheimer's diseased brain tissue panels are designed for rapidly screening Alzheimer's disease-specific genes or proteins in brain tissues. Tissues are arrayed on positively charged glass slides. Alzheimer's diseased brain tissues on T8236444Alz are selected based on functional regions, while those on T8236445Alz are selected based on anatomical regions. T8236564Alz contains both functional and anatomical regions of Alzheimer's diseased brain tissues. Normal brain tissue panels are also available in the identical format as controls. T8236446Alz contains both Alzheimer's diseased brain tissues and normal brain tissues.
Catalog No. 50-182-522 Supplier Biochain Institute Inc Supplier No. T8236564ALZ
BioChain's FFPE Alzheimer's diseased brain tissue panels are designed for rapidly screening Alzheimer's disease-specific genes or proteins in brain tissues. Tissues are arrayed on positively charged glass slides. Alzheimer's diseased brain tissues on T8236444Alz are selected based on functional regions, while those on T8236445Alz are selected based on anatomical regions. T8236564Alz contains both functional and anatomical regions of Alzheimer's diseased brain tissues. Normal brain tissue panels are also available in the identical format as controls. T8236446Alz contains both Alzheimer's diseased brain tissues and normal brain tissues.