Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Human Tumor, Same Tumor Type, Different Donors (5 tissues), 5 slides/PK
SDP
Supplier:  Biochain Institute Inc T8235142
Encompass_Preferred
BioChain's FFPE multiple tissue panels from the same type of tumors are designed for rapidly screening expression of tumor specific genes or proteins. Tumor tissue sections with the same tissue type collected from three different donors are arrayed along with a corresponding normal control tissue section on positively charged glass slides. Documentation of tissues' clinical histories may be available upon request. Human placenta tissue section is included in each panel as an interpanel control for comparison between different tissue panels. he tissue sections are approximately 5-10 um thick.
Catalog No. 50-182-541